Product Details

SNP ID
rs3174607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125529561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTTATTTTAATTTTTAACAGA[A/G]GTAAAATAAAGCATACTTATCCAAA
Phenotype
MIM: 606739 MIM: 605931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL11 PubMed Links

Gene Details

Gene
OSBPL11
Gene Name
oxysterol binding protein like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022776.4 3935 UTR 3 NP_073613.2
Gene
SNX4
Gene Name
sorting nexin 4
There are no transcripts associated with this gene.

View Full Product Details