Product Details

SNP ID
rs3205190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8603728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAAACATACAGCGCATGATTGG[A/C]TATTAATGCTTTGGAGAGATGGTTT
Phenotype
MIM: 605257
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AICDA PubMed Links

Gene Details

Gene
AICDA
Gene Name
activation-induced cytidine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520772.2 1321 UTR 3 XP_011519074.1
XM_011520773.2 1321 Intron XP_011519075.1

View Full Product Details