Product Details

SNP ID
rs3193251
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:26170806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCGTGCCAACCCCTTTTGCCT[C/T]TGGGTTTGGGGAGCGGAGGGCCTCT
Phenotype
MIM: 603272 MIM: 616698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNKSR1 PubMed Links

Gene Details

Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
There are no transcripts associated with this gene.

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
There are no transcripts associated with this gene.

Gene
ZNF593
Gene Name
zinc finger protein 593
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015871.4 582 UTR 3 NP_056955.2
XM_017001398.1 582 UTR 3 XP_016856887.1

View Full Product Details