Product Details

SNP ID
rs3733480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140862836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAGCATTTACTTGGGGACAGCA[C/T]GCACATTTTGTTAAGCACAGAAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF150 PubMed Links

Gene Details

Gene
RNF150
Gene Name
ring finger protein 150
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020724.1 6280 Intron NP_065775.1
XM_005263150.4 6280 UTR 3 XP_005263207.1
XM_011532147.2 6280 UTR 3 XP_011530449.1
XM_011532148.2 6280 UTR 3 XP_011530450.1
XM_017008475.1 6280 UTR 3 XP_016863964.1
XM_017008476.1 6280 UTR 3 XP_016863965.1

View Full Product Details