Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301073.1 | 32 | Intron | NP_001288002.1 | ||
NM_001301074.1 | 32 | Intron | NP_001288003.1 | ||
NM_001301075.1 | 32 | Intron | NP_001288004.1 | ||
NM_001301077.1 | 32 | Intron | NP_001288006.1 | ||
NM_001301080.1 | 32 | Intron | NP_001288009.1 | ||
NM_001301082.1 | 32 | Intron | NP_001288011.1 | ||
NM_001301083.1 | 32 | Intron | NP_001288012.1 | ||
NM_001301086.1 | 32 | Intron | NP_001288015.1 | ||
NM_004779.5 | 32 | Intron | NP_004770.4 | ||
XM_005268527.2 | 32 | Intron | XP_005268584.1 | ||
XM_011537706.1 | 32 | Intron | XP_011536008.1 | ||
XM_011537708.2 | 32 | Intron | XP_011536010.1 | ||
XM_017010048.1 | 32 | Intron | XP_016865537.1 | ||
XM_017010049.1 | 32 | Intron | XP_016865538.1 | ||
XM_017010050.1 | 32 | Intron | XP_016865539.1 | ||
XM_017010051.1 | 32 | Intron | XP_016865540.1 | ||
XM_017010052.1 | 32 | Intron | XP_016865541.1 | ||
XM_017010053.1 | 32 | Intron | XP_016865542.1 | ||
XM_017010054.1 | 32 | Intron | XP_016865543.1 | ||
XM_017010055.1 | 32 | Intron | XP_016865544.1 | ||
XM_017010056.1 | 32 | UTR 5 | XP_016865545.1 | ||
XM_017010057.1 | 32 | Intron | XP_016865546.1 | ||
XM_017010058.1 | 32 | Intron | XP_016865547.1 |