Product Details

SNP ID
rs3768334
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:26791313 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CATTGTGTCAGTTGAGTGGGAGAAA[C/T]GTGGCAATGTGCAAGTCCTAAAACC
Phenotype
MIM: 603024 MIM: 610274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID1A PubMed Links

Gene Details

Gene
ARID1A
Gene Name
AT-rich interaction domain 1A
There are no transcripts associated with this gene.

Gene
PIGV
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202554.1 Intron NP_001189483.1
NM_017837.3 Intron NP_060307.2

View Full Product Details