Product Details

SNP ID
rs3790441
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:38886820 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TAAAGGTCTAGAGAACTAGGGGTAA[A/G]GCAAAAGAGACACAAAACTCCATGT
Phenotype
MIM: 611923 MIM: 608962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJA9 PubMed Links

Gene Details

Gene
GJA9
Gene Name
gap junction protein alpha 9
There are no transcripts associated with this gene.

Gene
GJA9-MYCBP
Gene Name
GJA9-MYCBP readthrough
There are no transcripts associated with this gene.

Gene
RHBDL2
Gene Name
rhomboid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304746.1 Intron NP_001291675.1
NM_017821.4 Intron NP_060291.2

View Full Product Details