Product Details

SNP ID
rs3792717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106327709 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGACTCAGATTTATCAATAAAAT[A/G]GGGGCATGGATAGAACCTACTTCTT
Phenotype
MIM: 603605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIMP1 PubMed Links

Gene Details

Gene
AIMP1
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142415.1 Intron NP_001135887.1
NM_001142416.1 Intron NP_001135888.1
NM_004757.3 Intron NP_004748.2
XM_017008835.1 Intron XP_016864324.1
XM_017008836.1 Intron XP_016864325.1

View Full Product Details