Product Details

SNP ID
rs3211030
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:54032141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTATTACACTTGTCTTTTAAAA[A/G]AAAAATGCTTAAATTTCAGAATGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 904 UTR 3 NP_001291972.1
NM_001305049.1 904 UTR 3 NP_001291978.1
NM_001305050.1 904 UTR 3 NP_001291979.1
NM_001305051.1 904 UTR 3 NP_001291980.1
NM_001305052.1 904 UTR 3 NP_001291981.1
NM_001305066.1 904 Intron NP_001291995.1
NM_004872.4 904 UTR 3 NP_004863.2
XM_006711051.1 904 UTR 3 XP_006711114.1

View Full Product Details