Product Details

SNP ID
rs3792713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:168222255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGAGGATTATACATCACAAGAAA[A/G]GAATTCAATTGAACAACTCTAACAA
Phenotype
MIM: 613974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX60 PubMed Links

Gene Details

Gene
DDX60
Gene Name
DEXD/H-box helicase 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017631.5 Intron NP_060101.3
XM_011532103.2 Intron XP_011530405.1
XM_011532104.2 Intron XP_011530406.1
XM_017008383.1 Intron XP_016863872.1
XM_017008384.1 Intron XP_016863873.1

View Full Product Details