Product Details

SNP ID
rs3749828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133080448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATGCTCTCTCGCGCGGGGTCG[C/T]GGTTGTTTCTGGGAAGTGTGGTGGC
Phenotype
MIM: 604082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300776.1 246 UTR 5 NP_001287705.1
NM_001300777.1 246 UTR 5 NP_001287706.1
NM_001300778.1 246 UTR 5 NP_001287707.1
NM_003440.3 246 UTR 5 NP_003431.2

View Full Product Details