Product Details
- SNP ID
-
rs3750014
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:4775972 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGGGTCCCCGGAGGAGGTGACAC[T/C]TGAGCAGAGTCTTGGTGAGTGAAAG
- Phenotype
-
MIM: 613653
MIM: 616302
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AP5Z1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3750015] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AP5Z1
- Gene Name
- adaptor related protein complex 5 zeta 1 subunit
- Gene
- FOXK1
- Gene Name
- forkhead box K1
There are no transcripts associated with this gene.
View Full Product Details