Product Details

SNP ID
rs3813298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:118841378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTATACATTATTCAAACACCAC[A/G]AAAAATTGTTGCCATTATAATTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTWD2 PubMed Links

Gene Details

Gene
DTWD2
Gene Name
DTW domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308081.1 Intron NP_001295010.1
NM_173666.3 Intron NP_775937.1
XM_011543338.2 Intron XP_011541640.2
XM_011543340.2 Intron XP_011541642.1

View Full Product Details