Product Details

SNP ID
rs3814518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32541537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGGTGCTCTCTTCTCTTACATC[A/G]AGAGTTTGTTCAACTGAAGCTGCCA
Phenotype
MIM: 609507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOPORS PubMed Links

Gene Details

Gene
TOPORS
Gene Name
TOP1 binding arginine/serine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195622.1 2985 Silent Mutation CTC,CTT L931L NP_001182551.1
NM_005802.4 2985 Silent Mutation CTC,CTT L996L NP_005793.2
Gene
TOPORS-AS1
Gene Name
TOPORS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details