Product Details

SNP ID
rs3213483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32758800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGGAAGGATCAGCGGGAGCTCT[G/T]CCCTTTGTCTTGTGGGGCCCACAGT
Phenotype
MIM: 615161
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DQB2 PubMed Links

Gene Details

Gene
HLA-DQB2
Gene Name
major histocompatibility complex, class II, DQ beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198858.1 Intron NP_001185787.1
NM_001300790.1 Intron NP_001287719.1
XM_005249051.4 Intron XP_005249108.1
XM_011514560.2 Intron XP_011512862.1
XM_011514561.2 Intron XP_011512863.1
Gene
MIR3135B
Gene Name
microRNA 3135b
There are no transcripts associated with this gene.

View Full Product Details