Product Details

SNP ID
rs3742858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67471122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTAGATGGAGCAGAAACACAATG[C/T]TGGATGTTGCCAGGCACACAGGCAT
Phenotype
MIM: 603930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPHN PubMed Links

Gene Details

Gene
GPHN
Gene Name
gephyrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024218.1 3218 Intron NP_001019389.1
NM_020806.4 3218 Intron NP_065857.1
XM_005267254.3 3218 Intron XP_005267311.1
XM_011536340.2 3218 Intron XP_011534642.1
XM_011536342.2 3218 Intron XP_011534644.1
XM_011536343.2 3218 Intron XP_011534645.1
XM_011536344.2 3218 Intron XP_011534646.1
XM_011536345.2 3218 Intron XP_011534647.1
XM_011536346.2 3218 Intron XP_011534648.1
XM_011536347.2 3218 Intron XP_011534649.1
XM_017020913.1 3218 Intron XP_016876402.1
XM_017020914.1 3218 Intron XP_016876403.1
XM_017020915.1 3218 Intron XP_016876404.1
XM_017020916.1 3218 Intron XP_016876405.1
XM_017020917.1 3218 Intron XP_016876406.1
XM_017020918.1 3218 Intron XP_016876407.1
XM_017020919.1 3218 Intron XP_016876408.1
XM_017020920.1 3218 Intron XP_016876409.1
XM_017020921.1 3218 Intron XP_016876410.1
XM_017020922.1 3218 Intron XP_016876411.1
XM_017020923.1 3218 Intron XP_016876412.1
XM_017020924.1 3218 Intron XP_016876413.1
XM_017020925.1 3218 Intron XP_016876414.1
XM_017020926.1 3218 Intron XP_016876415.1
Gene
TMEM229B
Gene Name
transmembrane protein 229B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182526.2 3218 UTR 3 NP_872332.1
XM_005267373.2 3218 UTR 3 XP_005267430.1
XM_005267374.3 3218 UTR 3 XP_005267431.1
XM_005267375.2 3218 UTR 3 XP_005267432.1
XM_006720062.2 3218 UTR 3 XP_006720125.1
XM_017021051.1 3218 UTR 3 XP_016876540.1
XM_017021052.1 3218 UTR 3 XP_016876541.1
XM_017021053.1 3218 UTR 3 XP_016876542.1
XM_017021054.1 3218 UTR 3 XP_016876543.1
XM_017021055.1 3218 UTR 3 XP_016876544.1

View Full Product Details