Product Details

SNP ID
rs3744324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49291040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCATTTTCAGAGGCTACATATA[C/T]GCCCTTGGGGAGACAACTTAAGAAG
Phenotype
MIM: 613907
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6129 PubMed Links

Gene Details

Gene
MIR6129
Gene Name
microRNA 6129
There are no transcripts associated with this gene.

Gene
ZNF652
Gene Name
zinc finger protein 652
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145365.1 9513 UTR 3 NP_001138837.1
NM_014897.2 9513 UTR 3 NP_055712.1
XM_005257166.2 9513 UTR 3 XP_005257223.1

View Full Product Details