Product Details

SNP ID
rs3789683
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:94530506 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTTCTCCTTCCAGCTCTGCCCCACT[C/T]CTGCCTTTCTACACTTGTGTAGAGA
Phenotype
MIM: 134390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 1063 Missense Mutation AAG,GAG K228E NP_001171567.1
NM_001993.4 1063 Missense Mutation GAA,GGA E281G NP_001984.1

View Full Product Details