Product Details
- SNP ID
-
rs3802502
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:134031904 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGCCTGGTTTCAGTCATTTCC[A/G]GACTAACTGTGACAACGCGTGAGCA
- Phenotype
-
MIM: 601541
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BRD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114433245] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BRD3
- Gene Name
- bromodomain containing 3
- Gene
- LINC00094
- Gene Name
- long intergenic non-protein coding RNA 94
There are no transcripts associated with this gene.
View Full Product Details