Product Details

SNP ID
rs61751308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5949145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCGTGTCGGAGAGCAGCAGGAG[T/C]ACTGGTCCTGCACATCGTTGATGTC
Phenotype
MIM: 610109 MIM: 613160
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ANO2 PubMed Links

Gene Details

Gene
ANO2
Gene Name
anoctamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278596.1 8567 Intron NP_001265525.1
NM_001278597.1 8567 Intron NP_001265526.1
XM_006718988.3 8567 Intron XP_006719051.1
XM_011520975.2 8567 Intron XP_011519277.1
XM_011520978.2 8567 Intron XP_011519280.1
XM_017019672.1 8567 Intron XP_016875161.1
Gene
VWF
Gene Name
von Willebrand factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000552.4 8567 Missense Mutation TAC,TGC Y2771C NP_000543.2

View Full Product Details