Product Details

SNP ID
rs113306710
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:151728656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGGGCCACCAGTAGTAGCAGT[C/T]CTGCGTACCAAGCCATCCTAACTTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RIIAD1 PubMed Links

Gene Details

Gene
RIIAD1
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144956.1 Intron NP_001138428.1

View Full Product Details