Product Details

SNP ID
hCV27833379
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100188970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGATTTATGAAAGCCAACTAA[G/T]ATTAGAAGCACAGATACACAATCCC
Phenotype
MIM: 600664 MIM: 611604
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CHUK PubMed Links

Gene Details

Gene
CHUK
Gene Name
conserved helix-loop-helix ubiquitous kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278.4 2927 UTR 3 NP_001269.3
NM_001320928.1 2927 UTR 3 NP_001307857.1
Gene
ERLIN1
Gene Name
ER lipid raft associated 1
There are no transcripts associated with this gene.

View Full Product Details