Product Details

SNP ID
rs761690138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77696488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAAAATGGCGGAACCGCCGAGC[C/T]CCGTGCACTGTGTCGCTGCCGCGGC
Phenotype
MIM: 616048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APTR PubMed Links

Gene Details

Gene
APTR
Gene Name
Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
RSBN1L
Gene Name
round spermatid basic protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198467.2 63 Missense Mutation CCC,TCC P7S NP_940869.2

View Full Product Details