Product Details

SNP ID
rs924607511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60678057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGAGGGGAACAGTGCAAGGG[G/T]CCGGAGGGCGGCCGGGGGCAGCGGT
Phenotype
MIM: 607470
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAS3 PubMed Links

Gene Details

Gene
BCAS3
Gene Name
BCAS3, microtubule associated cell migration factor
There are no transcripts associated with this gene.

View Full Product Details