Product Details

SNP ID
rs141052812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44258632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCACAGGGCAGGCCTGTCTAAAA[A/G]GCCCCAATGCACACAGACATGCATG
Phenotype
MIM: 140572 MIM: 604548 MIM: 610788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90AB1 PubMed Links

Gene Details

Gene
HSP90AB1
Gene Name
heat shock protein 90 alpha family class B member 1
There are no transcripts associated with this gene.

Gene
MIR4647
Gene Name
microRNA 4647
There are no transcripts associated with this gene.

Gene
NFKBIE
Gene Name
NFKB inhibitor epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004556.2 2115 UTR 3 NP_004547.2
Gene
SLC35B2
Gene Name
solute carrier family 35 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286509.1 2115 Intron NP_001273438.1
NM_001286510.1 2115 Intron NP_001273439.1
NM_001286511.1 2115 Intron NP_001273440.1
NM_001286512.1 2115 Intron NP_001273441.1
NM_001286513.1 2115 Intron NP_001273442.1
NM_001286517.1 2115 Intron NP_001273446.1
NM_001286519.1 2115 Intron NP_001273448.1
NM_001286520.1 2115 Intron NP_001273449.1
NM_178148.3 2115 Intron NP_835361.1

View Full Product Details