Product Details

SNP ID
rs971316667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133041071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAGTGAATCTCACAATTACTTT[A/G]TTTCTGATATTTCCTTATCTGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928597 PubMed Links

Gene Details

Gene
LOC101928597
Gene Name
uncharacterized LOC101928597
There are no transcripts associated with this gene.

Gene
ZNF84
Gene Name
zinc finger protein 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127372.2 Intron NP_001120844.1
NM_001289971.1 Intron NP_001276900.1
NM_001289972.1 Intron NP_001276901.1
NM_003428.5 Intron NP_003419.3

View Full Product Details