Product Details

SNP ID
rs184379055
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:109540367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCTTCCCTAGCTTAGGAGGGG[A/C]ACGCTCTTGTCTGAAGGAAGGCCAG
Phenotype
MIM: 139370 MIM: 606916
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
There are no transcripts associated with this gene.

Gene
GPR61
Gene Name
G protein-coupled receptor 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031936.4 Intron NP_114142.3
XM_017002438.1 Intron XP_016857927.1

View Full Product Details