Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040138.2 | 186 | Intron | NP_001035228.1 | ||
NM_016326.3 | 186 | Intron | NP_057410.1 | ||
NM_016951.3 | 186 | Intron | NP_058647.1 | ||
NM_181640.2 | 186 | Intron | NP_857591.1 | ||
NM_181641.2 | 186 | Intron | NP_857592.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001202509.1 | 186 | Intron | NP_001189438.1 | ||
NM_001204098.1 | 186 | Intron | NP_001191027.1 | ||
NM_001204099.1 | 186 | Intron | NP_001191028.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_052999.3 | 186 | Silent Mutation | CCA,CCG | P21P | NP_443725.3 |
NM_181268.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851785.2 |
NM_181269.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851786.1 |
NM_181270.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851787.1 |
NM_181271.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851788.1 |
NM_181272.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851789.1 |
NM_181283.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851800.1 |
NM_181296.2 | 186 | Silent Mutation | CCA,CCG | P21P | NP_851813.1 |