Product Details

SNP ID
rs556875380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50517211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCAGCCTTCAGGAACTCCACC[C/T]AGAACTAGAACAGAAGAGATGGGTG
Phenotype
MIM: 609835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCBPAP PubMed Links

Gene Details

Gene
MYCBPAP
Gene Name
MYCBP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032133.4 Intron NP_115509.4
XM_005257725.3 Intron XP_005257782.1
XM_005257726.2 Intron XP_005257783.1
XM_011525342.1 Intron XP_011523644.1
XM_011525343.1 Intron XP_011523645.1
XM_017025205.1 Intron XP_016880694.1

View Full Product Details