Product Details

SNP ID
rs115689527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48549196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTACAATTTTTTCCTTTTTT[C/T]CCCTCATATAAATACATAATGTAGG
Phenotype
MIM: 142967 MIM: 142966
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB-AS1 PubMed Links

Gene Details

Gene
HOXB-AS1
Gene Name
HOXB cluster antisense RNA 1
There are no transcripts associated with this gene.

Gene
HOXB2
Gene Name
homeobox B2
There are no transcripts associated with this gene.

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 12832 UTR 3 NP_002137.4
XM_005257277.3 12832 UTR 3 XP_005257334.1
XM_005257280.3 12832 UTR 3 XP_005257337.1
XM_005257282.3 12832 UTR 3 XP_005257339.1
XM_006721854.2 12832 UTR 3 XP_006721917.1
XM_011524707.2 12832 UTR 3 XP_011523009.1
XM_011524708.2 12832 UTR 3 XP_011523010.1
XM_011524710.1 12832 UTR 3 XP_011523012.1
XM_011524719.1 12832 UTR 3 XP_011523021.1
XM_011524720.2 12832 UTR 3 XP_011523022.1
XM_011524721.2 12832 UTR 3 XP_011523023.1
XM_011524726.2 12832 UTR 3 XP_011523028.1
XM_017024557.1 12832 UTR 3 XP_016880046.1
XM_017024558.1 12832 UTR 3 XP_016880047.1
XM_017024559.1 12832 UTR 3 XP_016880048.1
XM_017024560.1 12832 UTR 3 XP_016880049.1
XM_017024561.1 12832 UTR 3 XP_016880050.1
XM_017024562.1 12832 UTR 3 XP_016880051.1
XM_017024563.1 12832 UTR 3 XP_016880052.1

View Full Product Details