Product Details
- SNP ID
-
rs4849068
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112488084 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAGCCTTGGTGGGGTTCAGTGCA[A/G]GACCCTGTGCAGCCCAGTCCTTCCC
- Phenotype
-
MIM: 608291
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TTL
PubMed Links
Gene Details
- Gene
- TTL
- Gene Name
- tubulin tyrosine ligase
View Full Product Details