Product Details

SNP ID
rs1860126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56537147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCATGACACCAGGGAGTGATTGC[C/T]TTCAGCAAACCTTCTGGTGGCTGGA
Phenotype
MIM: 616798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP28 PubMed Links

Gene Details

Gene
ZFP28
Gene Name
ZFP28 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308440.1 357 Intron NP_001295369.1
NM_020828.1 357 Intron NP_065879.1
XM_011526462.2 357 Intron XP_011524764.1
XM_011526463.2 357 UTR 5 XP_011524765.2
Gene
ZNF471
Gene Name
zinc finger protein 471
There are no transcripts associated with this gene.

View Full Product Details