Product Details

SNP ID
rs4493312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69321741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAAAAACACATTCAACCACAGA[A/T]CATTCAGAAAGCTAACAGGATCATT
Phenotype
MIM: 138292
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GFPT1 PubMed Links

Gene Details

Gene
GFPT1
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244710.1 6677 UTR 3 NP_001231639.1
NM_002056.3 6677 UTR 3 NP_002047.2

View Full Product Details