Product Details

SNP ID
hCV27907201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24141285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTATGAGATATTCGTTACTTT[A/T]TAATAGAAGCTGTGCCCTTCTATAA
Phenotype
MIM: 612135
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CABYR PubMed Links

Gene Details

Gene
CABYR
Gene Name
calcium binding tyrosine phosphorylation regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308231.1 Intron NP_001295160.1
NM_012189.3 Intron NP_036321.2
NM_138643.2 Intron NP_619584.1
NM_138644.2 Intron NP_619585.1
NM_153768.2 Intron NP_722452.1
NM_153769.2 Intron NP_722453.1
NM_153770.2 Intron NP_722454.1
Gene
TTC39C
Gene Name
tetratricopeptide repeat domain 39C
There are no transcripts associated with this gene.

View Full Product Details