Product Details

SNP ID
rs4966710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46899505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTCACAGGGCAGTGAGGGAGA[A/G]GAGCTGAGGCTGGCCCTGGTCTGAG
Phenotype
MIM: 138210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPT2 PubMed Links

Gene Details

Gene
GPT2
Gene Name
glutamic--pyruvic transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142466.2 Intron NP_001135938.1
NM_133443.3 Intron NP_597700.1
XM_017023790.1 Intron XP_016879279.1

View Full Product Details