Product Details

SNP ID
rs4752442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:120598830 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGGGGGAGGGGTAGGAGTGGGA[C/G]ACACAAGTGTGGTTTGGGACAAAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01561 PubMed Links

Gene Details

Gene
LINC01561
Gene Name
long intergenic non-protein coding RNA 1561
There are no transcripts associated with this gene.

Gene
PLPP4
Gene Name
phospholipid phosphatase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030059.2 10165 Intron NP_001025230.1
NM_001318166.1 10165 Intron NP_001305095.1
NM_001318167.1 10165 Intron NP_001305096.1
NM_001318168.1 10165 Intron NP_001305097.1
NM_001318169.1 10165 Intron NP_001305098.1
XM_005269592.2 10165 UTR 3 XP_005269649.1
XM_006717685.3 10165 UTR 3 XP_006717748.1
XM_006717686.3 10165 Intron XP_006717749.1
XM_011539444.2 10165 UTR 3 XP_011537746.1
XM_011539445.2 10165 Intron XP_011537747.1
XM_017015820.1 10165 Intron XP_016871309.1
XM_017015821.1 10165 UTR 3 XP_016871310.1
XM_017015822.1 10165 UTR 3 XP_016871311.1
XM_017015823.1 10165 Intron XP_016871312.1
XM_017015824.1 10165 Intron XP_016871313.1

View Full Product Details