Product Details

SNP ID
rs4474637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64747520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTAGTGCTCATCACCTGGCTGGT[C/T]AAAGGTTGAGGTGGGCTCAAAAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBPMS2 PubMed Links

Gene Details

Gene
RBPMS2
Gene Name
RNA binding protein with multiple splicing 2
There are no transcripts associated with this gene.

View Full Product Details