Product Details
- SNP ID
-
rs4638523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:62643822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGAACCCAGACTTAGCCACTCAC[C/G]TTCTCCGCAATAGACCCGGGCAGCC
- Phenotype
-
MIM: 607349
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MGC15885
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74934048] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGC15885
- Gene Name
- uncharacterized protein MGC15885
There are no transcripts associated with this gene.
- Gene
- TLN2
- Gene Name
- talin 2
View Full Product Details