Product Details

SNP ID
rs4766147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:3616801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTTGCAATCCCATACAAAGGCC[C/T]AGGACCCCGTTGCCTATACTCAGAG
Phenotype
MIM: 614178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRACR2A PubMed Links

Gene Details

Gene
CRACR2A
Gene Name
calcium release activated channel regulator 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144958.1 Intron NP_001138430.1
NM_032680.3 Intron NP_116069.1
XM_006719021.3 Intron XP_006719084.1
XM_011521034.2 Intron XP_011519336.1
XM_011521036.2 Intron XP_011519338.1
XM_011521037.2 Intron XP_011519339.1
XM_011521038.2 Intron XP_011519340.1

View Full Product Details