Product Details

SNP ID
rs4735020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:107905363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAAAGAGATTAAAGCTTCTCTCT[A/C]TATATATATGTATACGTAATACATG
Phenotype
MIM: 610575
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RSPO2 PubMed Links

Gene Details

Gene
RSPO2
Gene Name
R-spondin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282863.1 Intron NP_001269792.1
NM_001317942.1 Intron NP_001304871.1
NM_178565.4 Intron NP_848660.3
XM_011517018.1 Intron XP_011515320.1
XM_011517019.1 Intron XP_011515321.1
XM_017013395.1 Intron XP_016868884.1

View Full Product Details