Product Details

SNP ID
rs4771983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:97146479 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATGAATGAAAAATGAGCACAGAA[A/G]GAGTGCTGTTGGCCAGGGATCTCAC
Phenotype
MIM: 607327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBNL2 PubMed Links

Gene Details

Gene
MBNL2
Gene Name
muscleblind like splicing regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306070.1 Intron NP_001292999.1
NM_144778.3 Intron NP_659002.1
NM_207304.2 Intron NP_997187.1
XM_005254018.2 Intron XP_005254075.1
XM_005254019.2 Intron XP_005254076.1
XM_005254021.2 Intron XP_005254078.1
XM_005254022.2 Intron XP_005254079.1
XM_006719912.2 Intron XP_006719975.1
XM_011521045.2 Intron XP_011519347.1
XM_017020308.1 Intron XP_016875797.1
XM_017020309.1 Intron XP_016875798.1
XM_017020310.1 Intron XP_016875799.1
XM_017020311.1 Intron XP_016875800.1

View Full Product Details