Product Details

SNP ID
rs4978600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114608258 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACTGAGCATTTTCTTCTAAGCA[C/T]TTCATCACTTCAATTAATTCTTGCA
Phenotype
MIM: 607296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1G1 PubMed Links

Gene Details

Gene
ATP6V1G1
Gene Name
ATPase H+ transporting V1 subunit G1
There are no transcripts associated with this gene.

Gene
TMEM268
Gene Name
transmembrane protein 268
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153045.3 Intron NP_694590.2
XM_006716997.1 Intron XP_006717060.1
XM_011518350.1 Intron XP_011516652.1
XM_011518351.2 Intron XP_011516653.1
XM_011518352.1 Intron XP_011516654.1
XM_011518353.1 Intron XP_011516655.1
XM_011518354.1 Intron XP_011516656.1
XM_011518355.1 Intron XP_011516657.1
XM_017014429.1 Intron XP_016869918.1
XM_017014430.1 Intron XP_016869919.1

View Full Product Details