Product Details
- SNP ID
-
rs7024579
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:136208567 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCGTGTTCTTCCCTTGTTAGAAGAG[C/T]GTTTGTAAAACCAGGACTTTGAAGC
- Phenotype
-
MIM: 600577
MIM: 612860
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LHX3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111392257] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LHX3
- Gene Name
- LIM homeobox 3
There are no transcripts associated with this gene.
- Gene
- QSOX2
- Gene Name
- quiescin sulfhydryl oxidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181701.3 |
2296 |
UTR 3 |
|
|
NP_859052.3 |
View Full Product Details