Product Details

SNP ID
rs7564925
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43806728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATTAAAGGATTAGGGTATTTGT[C/G]AGCATCATGAGTTTTCACATTCAAA
Phenotype
MIM: 605459
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ABCG5 PubMed Links

Gene Details

Gene
ABCG5
Gene Name
ATP binding cassette subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022436.2 Intron NP_071881.1
XM_005264480.3 Intron XP_005264537.1
XM_006712073.3 Intron XP_006712136.1
XM_006712074.3 Intron XP_006712137.1
XM_011533024.2 Intron XP_011531326.1
XM_011533025.2 Intron XP_011531327.1
XM_011533026.2 Intron XP_011531328.1
XM_011533027.2 Intron XP_011531329.1
XM_011533028.2 Intron XP_011531330.1
Gene
DYNC2LI1
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193464.1 Intron NP_001180393.1
NM_015522.3 Intron NP_056337.1
NM_016008.3 Intron NP_057092.2
XM_005264364.4 Intron XP_005264421.1
XM_005264365.4 Intron XP_005264422.1

View Full Product Details