Product Details

SNP ID
rs7815715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10527189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTTTCCTGTGTTGTTAAAAAT[C/G]CCCCATGAGCCTAACTTTGTGGCTG
Phenotype
MIM: 615144
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS55 PubMed Links

Gene Details

Gene
PRSS55
Gene Name
protease, serine 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197020.1 Intron NP_001183949.1
NM_198464.3 Intron NP_940866.2
XM_005272372.3 Intron XP_005272429.2
XM_011543814.2 Intron XP_011542116.2
XM_017013182.1 Intron XP_016868671.1

View Full Product Details