Product Details

SNP ID
rs8091400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74316590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCTCCCTCACGTGTAGAATTGC[A/G]TAGGTGCCTGTTGCCTGCTCCTAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C18orf63 PubMed Links

Gene Details

Gene
C18orf63
Gene Name
chromosome 18 open reading frame 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174123.1 Intron NP_001167594.1

View Full Product Details