Product Details

SNP ID
rs7114921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47661469 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGGTGATTGGACTATCAGTCTT[C/G]TCACTCCTAGAGGATCTTGTCTTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGBL2 PubMed Links

Gene Details

Gene
AGBL2
Gene Name
ATP/GTP binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024783.3 3235 Intron NP_079059.2
XM_005253138.4 3235 Intron XP_005253195.1
XM_005253139.4 3235 Intron XP_005253196.1
XM_005253140.4 3235 Intron XP_005253197.1
XM_005253141.4 3235 Intron XP_005253198.1
XM_006718327.3 3235 Intron XP_006718390.1
XM_006718328.3 3235 Intron XP_006718391.1
XM_011520372.2 3235 Intron XP_011518674.1
XM_011520373.2 3235 Intron XP_011518675.1
XM_011520374.2 3235 Intron XP_011518676.1
XM_011520375.2 3235 Intron XP_011518677.1
XM_011520376.2 3235 Intron XP_011518678.1
XM_011520377.2 3235 Intron XP_011518679.1
XM_011520378.2 3235 Intron XP_011518680.1
XM_011520379.2 3235 Intron XP_011518681.1
XM_011520381.2 3235 Intron XP_011518683.1
XM_017018331.1 3235 UTR 3 XP_016873820.1
XM_017018332.1 3235 Intron XP_016873821.1
XM_017018333.1 3235 Intron XP_016873822.1

View Full Product Details