Product Details

SNP ID
rs7129565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63974937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGTTTGGGCATGTCTGGAGGGC[G/T]CGAACGACGGGGACAGTGCCCGGGC
Phenotype
MIM: 123870
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COX8A PubMed Links

Gene Details

Gene
COX8A
Gene Name
cytochrome c oxidase subunit 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004074.2 Intron NP_004065.1

View Full Product Details