Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015059.2 | Intron | NP_055874.2 | |||
XM_005254708.4 | Intron | XP_005254765.1 | |||
XM_005254710.4 | Intron | XP_005254767.1 | |||
XM_005254711.4 | Intron | XP_005254768.1 | |||
XM_005254712.4 | Intron | XP_005254769.1 | |||
XM_005254713.4 | Intron | XP_005254770.1 | |||
XM_005254714.3 | Intron | XP_005254771.1 | |||
XM_005254715.2 | Intron | XP_005254772.1 | |||
XM_006720717.3 | Intron | XP_006720780.1 | |||
XM_017022665.1 | Intron | XP_016878154.1 | |||
XM_017022666.1 | Intron | XP_016878155.1 | |||
XM_017022667.1 | Intron | XP_016878156.1 | |||
XM_017022668.1 | Intron | XP_016878157.1 | |||
XM_017022669.1 | Intron | XP_016878158.1 |