Product Details

SNP ID
rs4900314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96212866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGGTAGGATAGCAGGAGTTAA[C/T]GCCCCCTCCCCCATTCCATGGCCCC
Phenotype
MIM: 113503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BDKRB2 PubMed Links

Gene Details

Gene
BDKRB2
Gene Name
bradykinin receptor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000623.3 Intron NP_000614.1

View Full Product Details